missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial ribonuclease P protein 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17295
This item is not returnable.
View return policy
Description
Mitochondrial ribonuclease P protein 3 Polyclonal specifically detects Mitochondrial ribonuclease P protein 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Mitochondrial ribonuclease P protein 3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| KIAA0391, mitochondrial ribonuclease P protein 3, MRPP3Mitochondrial RNase P protein 3 | |
| Rabbit | |
| 67 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| O15091 | |
| KIAA0391 | |
| Recombinant protein encompassing a sequence within the center region of human KIAA0391. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9692 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction