missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondrial ribosomal protein L11 Antibody (OTI1E5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72707C
This item is not returnable.
View return policy
Description
Mitochondrial ribosomal protein L11 Monoclonal specifically detects Mitochondrial ribosomal protein L11 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Mitochondrial ribosomal protein L11 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MRPL11 | |
| Full length recombinant protein of human Mitochondrial ribosomal protein L11 (NP_057134) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E5 | |
| Western Blot, Immunohistochemistry | |
| L11MT, MGC111024, mitochondrial ribosomal protein L11, MRP-L11,39S ribosomal protein L11, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 65003.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction