missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitocondrial Translational Initiation Factor 3 Antibody (OTI4B6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72708AF405
This item is not returnable.
View return policy
Description
Mitocondrial Translational Initiation Factor 3 Monoclonal specifically detects Mitocondrial Translational Initiation Factor 3 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Mitocondrial Translational Initiation Factor 3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MTIF3 | |
| Recombinant protein fragment corresponding to amino acids 56-278 of human Mitocondrial Translational Initiation Factor 3 (NP_690876) produced in E.coli. | |
| 0.1 mL | |
| 219402.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4B6 | |
| Western Blot, Immunohistochemistry | |
| FLJ33676, IF3(mt), IF-3mt, mitochondrial, mitochondrial translational initiation factor 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction