Learn More
Mitofusin 1 Antibody (11E9-1H12), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-71775AF750
Product Code. 18226504
  • € 602.00 / 0.10mL
Description
Mitofusin 1 Monoclonal specifically detects Mitofusin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mitofusin 1
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
MFN1
Mouse Mitofusin 1 [Swiss-Prot: Q811U4]
Primary
Human, Mouse, Rat
IgG2b κ
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
11E9-1H12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1
Mouse
0.1 mL
55669
Store at 4°C in the dark.