missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitofusin 1 Antibody (11G8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Mitofusin 1 |
|---|---|
| Clone | 11G8 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30039876
|
Novus Biologicals
NBP3-26143-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30027983
|
Novus Biologicals
NBP3-26143-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mitofusin 1 Monoclonal antibody specifically detects Mitofusin 1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Mitofusin 1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1 | |
| A synthesized peptide derived from Human Mitofusin 1 [UniProt Q8IWA4] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 11G8 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Mitochondrial Fusion Proteins, Mitochondrial Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 55669 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title