missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mitofusin 1 Recombinant Protein Antigen

Product Code. 18231125 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18231125 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18231125 Supplier Novus Biologicals™ Supplier No. NBP257125PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Mitofusin 1. Source: E.coli Amino Acid Sequence: SLGWSSLVHRFLGPRNAQRVLLGLSEPIFQLPRSLASTPTAPTTPATPDNASQEELMITLVTGL The Mitofusin 1 Recombinant Protein Antigen is derived from E. coli. The Mitofusin 1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55669
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name Mitofusin 1 Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1
Gene Symbol MFN1
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-52335. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.