Learn More
Mitomycin A, TRC
Product Code. 30004582
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004582 10 mg 10mg
30009060 25 mg 25mg
30007923 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004582 Supplier Toronto Research Chemicals Supplier No. TRCM37189010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4055-39-4
Molecular Formula C16 H19 N3 O6
Molecular Weight (g/mol) 349.34
Synonym Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, (1aS,8S,8aR,8bS)-, Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 1,1a,2,8,8a,8b-hexahydro-8-(hydroxymethyl)-6,8a-dimethoxy-5-methyl-, carbamate (ester) (8CI), Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, [1aS-(1aα,8β,8aα,8bα)]-, Mitomycin A (7CI), (1aS,8S,8aR,8bS)-8-[[(Aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, NSC 75986, Mitomycin Imp. B (EP)
SMILES COC1=C(C)C(=O)C2=C([C@@H](COC(=O)N)[C@@]3(OC)[C@H]4N[C@H]4CN23)C1=O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H19N3O6/c1-6-11(20)10-9(12(21)13(6)23-2)7(5-25-15(17)22)16(24-3)14-8(18-14)4-19(10)16/h7-8,14,18H,4-5H2,1-3H3,(H2,17,22)/t7-,8+,14+,16-/m1/s1
Formula Weight 349.1274
Percent Purity >95%
Chemical Name or Material Mitomycin A
Grade HPLC