Learn More
Mixed Calibration Standard 1A For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep MIXSTD1A-100
Additional Details : Weight : 0.12500kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 100μg/mL |
Ca | 100μg/mL |
Sb | 50μg/mL |
Se | 50μg/mL |
B | 20μg/mL |
Cd | 20μg/mL |
Cu | 20μg/mL |
Mn | 20μg/mL |
Ba | 10μg/mL |
Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Mixed Calibration Standard 1A | |
125 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Components in 5% HNO3: Sb •50μg/mL As •100μg/mL Ba •10μg/mL B •10μg/mL Cd •20μg/mL Ca •100μg/mL Cu •20μg/mL Mn •20μg/mL Se •50μg/mL Ag •5μg/mL |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
5% HNO3/tr Tart. Acid/tr HF |
Product intended for laboratory use only