missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIXL1, Mouse anti-Human, Clone: 4D11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00083881-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: QRRKRTSFSAEQLQLLELVFRRTRYPDIHLRERLAALTLLPESRIQVWFQNRRAKSRRQSGKSFQPLARPEIILNHCAPGTETKCLKPQLPLEVDVNCLPSpecifications
| MIXL1 | |
| Monoclonal | |
| Unconjugated | |
| Mix1 homeobox-like 1 (Xenopus laevis) | |
| MGC138179/MILD1/MIX/MIXL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4D11 | |
| In 1x PBS, pH 7.4 | |
| NM_031944 | |
| MIXL1 | |
| MIXL1 (NP_114150, 86 a.a. ∼ 185 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 83881 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction