missing translation for 'onlineSavingsMsg'
Learn More

MK-4101, MedChemExpress

Product Code. 16499456
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499456

missing translation for 'mfr': Medchem Express HY100036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-4101 is a Smoothened (SMO) antagonist (IC50 of 1.1 μM for 293 cells ) and also a potent inhibitor of the hedgehog pathway (IC50 of 1.5 μM for mouse cells; IC50 of 1 μM for KYSE180 oesophageal cancer cells). MK-4101 has robust antitumor activity that inhibits tumor cell proliferation and induces extensive apoptosis.

MK-4101 is a Smoothened (SMO) antagonist (IC50 of 1.1 μM for 293 cells ) and also a potent inhibitor of the hedgehog pathway (IC50 of 1.5 μM for mouse cells

Specifications

Chemical Name or Material MK-4101
Physical Form Solid
CAS 935273-79-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H24F5N5O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (101.32 mM)
SMILES FC(C1=CC=CC=C1C2=NN=C(C34CCC(C5=NOC(C6CC(F)(F)C6)=N5)(CC4)CC3)N2C)(F)F
Molecular Weight (g/mol) 493.47
Formula Weight 493.47
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.