Learn More
MK-5108, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Specifications
Specifications
| Chemical Name or Material | MK-5108 |
| CAS | 1010085-13-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H21ClFN3O3S |
| Quantity | 50 mg |
| Synonym | VX-689 |
| Solubility Information | DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic) |
| SMILES | FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3 |
| Molecular Weight (g/mol) | 461.94 |
| Show More |
Research purposes only