Learn More
MK-5108, MedChemExpress
Product Code. 16438287
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16438287
Brand: Medchem Express HY13252
MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Specifications
Chemical Name or Material MK-5108
CAS 1010085-13-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClFN3O3S
Quantity 5 mg
Synonym VX-689
Solubility Information DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic)
SMILES FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3
Molecular Weight (g/mol) 461.94
Formula Weight 461.94
Percent Purity 96.69%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only