missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-8033 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
MK-8033 hydrochloride is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 hydrochloride can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).
Specifications
Specifications
| Chemical Name or Material | MK-8033 hydrochloride |
| CAS | 1283000-43-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H22ClN5O3S |
| Quantity | 5 mg |
| Solubility Information | H2O : 7.14 mg/mL (14.06 mM; Need ultrasonic) ∣DMSO : 5.88 mg/mL (11.58 mM; Need ultrasonic) |
| SMILES | O=S(CC1=CC=C2C(C(C3=CC(C4=CN(C)N=C4)=CN=C3C=C2)=O)=C1)(NCC5=NC=CC=C5)=O.Cl |
| Molecular Weight (g/mol) | 507.99 |
| Formula Weight | 507.99 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction