missing translation for 'onlineSavingsMsg'
Learn More

MK-8617, MedChemExpress

Product Code. 16402686
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16402686

Brand: Medchem Express HY101023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-8617 is an orally active pan-inhibitor of hypoxia-inducible factor prolyl hydroxylase 1-3 (HIF PHD1-3) with an IC50 of 1 nM for PHD2.

MK-8617 is an orally active pan-inhibitor of hypoxia-inducible factor prolyl hydroxylase 1-3 (HIF PHD1-3) with an IC50 of 1 nM for PHD2.

Specifications

Chemical Name or Material MK-8617
CAS 1187990-87-9
Color White
Purity Grade Notes Research
Molecular Formula C24H21N5O4
Quantity 25 mg
Solubility Information DMSO : 6 mg/mL (13.53 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(C2=NN=CC=C2)N=C1O)NC(C3=CC=C(OC)C=C3)C4=CC=C(OC)C=C4
Molecular Weight (g/mol) 443.45
Formula Weight 443.45
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.