Learn More
MK-8617, MedChemExpress
Product Code. 16494496
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
Product Code. 16494496
Brand: Medchem Express HY101023
MedChemExpress MK-8617 is an orally active pan-inhibitor of hypoxia-inducible factor prolyl hydroxylase 1-3 (HIF PHD1-3) with an IC50 of 1 nM for PHD2.
MK-8617 is an orally active pan-inhibitor of hypoxia-inducible factor prolyl hydroxylase 1-3 (HIF PHD1-3) with an IC50 of 1 nM for PHD2.
Specifications
Chemical Name or Material MK-8617
CAS 1187990-87-9
Color White
Purity Grade Notes Research
Molecular Formula C24H21N5O4
Quantity 100 mg
Solubility Information DMSO : 6 mg/mL (13.53 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(C2=NN=CC=C2)N=C1O)NC(C3=CC=C(OC)C=C3)C4=CC=C(OC)C=C4
Molecular Weight (g/mol) 443.45
Formula Weight 443.45
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only