missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mkl1 Antibody (OTI3G7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72715JF549
This item is not returnable.
View return policy
Description
Mkl1 Monoclonal specifically detects Mkl1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Mkl1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MRTFA | |
| Recombinant protein fragment corresponding to amino acids 165-397 of human Mkl1 (NP_065882) produced in E.coli. | |
| 0.1 mL | |
| 57591.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3G7 | |
| Western Blot, Immunohistochemistry | |
| AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction