missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKL2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-46208CL1
This item is not returnable.
View return policy
Description
MKL2 Polyclonal antibody specifically detects MKL2 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| MKL2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001 | |
| The immunogen for this product maps to a region between residue 900 and 950 of human Megakaryoblastic Leukemia 2 using the numbering given in entry NP_054767.3 (GeneID 57496). | |
| 0.1 mL | |
| Cardiovascular Biology, Stem Cells | |
| 57496 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur