missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKLP1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-211H
This item is not returnable.
View return policy
Description
MKLP1 Polyclonal antibody specifically detects MKLP1 in Human samples. It is validated for Western Blot
Specifications
| MKLP1 | |
| Polyclonal | |
| Western Blot | |
| kinesin family member 23, kinesin-like 5 (mitotic kinesin-like protein 1), Kinesin-like protein 5, kinesin-like protein KIF23, KNSL5CHO1, mitotic kinesin-like 1, MKLP-1, MKLP1Mitotic kinesin-like protein 1 | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Mitotic Kinesin-Like 1 using the numbering given in entry NP_004847.2 (GeneID 9493). | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 9493 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction