missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKLP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-211
This item is not returnable.
View return policy
Description
MKLP1 Polyclonal specifically detects MKLP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MKLP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| kinesin family member 23, kinesin-like 5 (mitotic kinesin-like protein 1), Kinesin-like protein 5, kinesin-like protein KIF23, KNSL5CHO1, mitotic kinesin-like 1, MKLP-1, MKLP1Mitotic kinesin-like protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1-10 ug/mg | |
| NP_004847.2 | |
| KIF23 | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Mitotic Kinesin-Like 1 using the numbering given in entry NP_004847.2 (GeneID 9493). | |
| 0.1 mg | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 9493 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction