Learn More
MKNK2B Polyclonal Antibody
GREENER_CHOICE
MKNK2B Polyclonal Antibody
Brand:  Invitrogen PA595641
Product Code. 16578481
  • € 578.00 / 100µL
Description
MKNK2B Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MKNK2B
Polyclonal
Unconjugated
MKNK2
G protein-coupled receptor kinase 7; GPRK7; MAP kinase interacting serine/threonine kinase 2; MAP kinase signal-integrating kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; MAPK signal-integrating kinase 2; MKNK2; MNK2
Rabbit
Antigen Affinity Chromatography
RUO
2872
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.15 mg/mL
10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Q9HBH9
MKNK2
Synthetic peptide from the C-terminal region, specific to the human Mnk2b isoform.