missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MKP3 Recombinant Rabbit Monoclonal Antibody (1V5L10)
GREENER_CHOICE

Product Code. 17155713
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17155713 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17155713 Supplier Invitrogen™ Supplier No. MA535048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ARGKNCGVLV HCLAGISRSV TVTVAYLMQK LNLSMNDAYD IVKMKKSNIS PNFNFMGQLL DFERTLGLSS PCDNRVPAQQ LYFTTPSNQN VYQVDSLQST.

DUSP6 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP)kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associatedwith cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK2, is expressed in a variety of tissues with the highest levels in heart and pancreas, and unlike most other members of this family, is localized in the cytoplasm. Twotranscript variants encoding different isoforms have been found for the DUSP6 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1V5L10
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene DUSP6
Gene Accession No. Q16828, Q64346, Q9DBB1
Gene Alias 1300019I03Rik; dual specificity phosphatase 6; dual specificity protein phosphatase 6; Dual specificity protein phosphatase PYST1; Dusp6; DUSP6-ALT; HH19; MAP kinase phosphatase 3; mitogen-activated protein kinase phosphatase 3; Mkp3; MKP-3; PYST1; serine/threonine specific protein phosphatase
Gene Symbols DUSP6
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116663, 1848, 67603
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.