missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN1 Antibody (OTI2C8), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72718MFV450
This item is not returnable.
View return policy
Description
MKRN1 Monoclonal antibody specifically detects MKRN1 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| MKRN1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| OTI2C8 | |
| Flow Cytometry | |
| E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61 | |
| Full length human recomnbinant protein of human MKRN1(NP_038474) produced in HEK293T cell. | |
| 0.1 mL | |
| Zinc Finger | |
| 23608 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction