missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN1 Antibody (OTI2C8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72718PCP
This item is not returnable.
View return policy
Description
MKRN1 Monoclonal specifically detects MKRN1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| MKRN1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| MKRN1 | |
| Full length recombinant protein of human MKRN1 (NP_038474) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Flow Cytometry | |
| OTI2C8 | |
| Flow Cytometry | |
| E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 23608.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction