missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55250H
This item is not returnable.
View return policy
Description
MKRN2 Polyclonal antibody specifically detects MKRN2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MKRN2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 6.3.2.-, HSPC070, makorin ring finger protein 2, probable E3 ubiquitin-protein ligase makorin-2, RNF62RING finger protein 62 | |
| The immunogen recognized by this antibody maps to a region between residue 366 and the C-terminus (residue 416) of human makorin, ring finger protein, 2 using the numbering given in entry NP_054879.3 (GeneID 23609). | |
| 0.1 mL | |
| Signal Transduction, Zinc Finger | |
| 23609 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?