Learn More
Invitrogen™ MKRN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588429
Description
Immunogen sequence: MSTKQITCRY FMHGVCREGS QCLFSHDLAN SKPSTICKYY QKGYCAYGTR CRYDHTRPSA AAGGAVGTMA HSVPSPAFHS PHPPSEVTAS IVKTNSHEPG KREKRTLVLR DRNLSGMAER KTQPSMVSNP GSCSDPQPSP EMKPHSYLDA IRSGLDDVEA SSSYSNEQQL CPYAAAGECR FGDACVYLHG EVCEICRLQV LHPFDPEQRK AHEKICMLTF EHEMEKAFAF QASQDKVCSI; Positive Samples: Mouse liver.
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
Specifications
| MKRN2 | |
| Polyclonal | |
| Unconjugated | |
| MKRN2 | |
| 2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23609 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9H000 | |
| MKRN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human MKRN2 (NP_0548793). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.