Learn More
MKRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12238-1-AP-150UL
 View more versions of this product
Product Code. 16849833
  • € 410.00 / 150µL
Description
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins.
Specifications
Specifications
MKRN2
Polyclonal
Unconjugated
MKRN2
HSPC070, makorin ring finger protein 2, MKRN2, RING finger protein 62, RNF62
Rabbit
Antigen Affinity Chromatography
RUO
23609, 297525, 67027
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9ERV1, Q9H000
Mkrn2
MKRN2 Fusion Protein Ag2881
150 μL
Primary
Human, Mouse, Rat