Learn More
MKRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12238-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins.Specifications
| MKRN2 | |
| Polyclonal | |
| Unconjugated | |
| MKRN2 | |
| HSPC070, makorin ring finger protein 2, MKRN2, RING finger protein 62, RNF62 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23609, 297525, 67027 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ERV1, Q9H000 | |
| Mkrn2 | |
| MKRN2 Fusion Protein Ag2881 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |