Learn More
MKX Mouse anti-Human, Clone: OTI6B9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808429
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MKX is a member of an Iroquois (IRX) family-related class of ′three-amino acid loop extension′ (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.Specifications
| MKX | |
| Monoclonal | |
| Unconjugated | |
| MKX | |
| C10orf48, IFRX, IRXL1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 283078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6B9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8IYA7 | |
| MKX | |
| Full length human recombit protein of human MKX produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.