missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540565
This item is not returnable.
View return policy
Description
Peptide sequence: IKSENSVIKA GVRPESRASE DYVAPPKYKS SLLNRYLNDS LRHVMATNTT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 92%; Zebrafish: 85%.
MKX is a member of an Iroquois (IRX) family-related class of 'three-amino acid loop extension' (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.
Specifications
| MKX | |
| Polyclonal | |
| Unconjugated | |
| MKX | |
| 9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IYA7 | |
| MKX | |
| Synthetic peptide directed towards the middle region of human MKX. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction