missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569056
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 83%.
MKX is a member of an Iroquois (IRX) family-related class of 'three-amino acid loop extension' (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.
Specifications
| MKX | |
| Polyclonal | |
| Unconjugated | |
| MKX | |
| 9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 210719 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BIA3 | |
| MKX | |
| synthetic peptide directed towards the middle region of human MKX (aa 290-338). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction