missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 382
GREENER_CHOICE

Product Code. 18701088
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18701088

Brand: Tocris Bioscience™ 5738/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MrgX1 (SNSR4) positive allosteric modulator

ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mas-Related G Protein Coupled Receptor Modulators
CAS 1646499-97-9
Quantity 10 mg
Target Sensory Neuron-Specific Receptor Agonists
Molecular Formula C18H20N2O4S
Formula Weight 360.43
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.