missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML T7

Product Code. 30042724
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30042724

Brand: Tocris Bioscience™ 8119/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of phosphatidylserine/CEACAM1 binding to Tim-3

ML T7 is a T cell immunoglobulin and mucin-containing molecule 3 (Tim-3) inhibitor; disrupts phosphatidylserine/CAECAM1 binding to Tim-3 (Kd = 7.0 μM and 7.4 μM for hTim-3 and mTim-3 respectively). ML T7 promotes cell proliferation, cytokine production and cytotoxicity of OT-I CD8+ cytotoxic T lymphocytes. ML T7 inhibits tumor growth in a syngeneic mouse model of hepatocellular carcinoma, increases NK cell tumor killing activity and DC maturation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(2,4-Dichlorophenyl)-3a,6a-dihydro-3-(4-methylphenyl)spiro[1H-furo[3,4-c]pyrrole-1,2'-[2H]indene]-1',3',4,6(3H,5H)-tetrone
CAS 459789-75-4
Quantity 10 mg
Target CEACAM1/CD66a
Molecular Formula C27H17Cl2NO5
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.