missing translation for 'onlineSavingsMsg'
Learn More

ML221, MedChemExpress

Product Code. 16410197
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410197

Brand: Medchem Express HY103254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML221 is a potent apelin (APJ) functional antagonist, inhibiting apelin-13-mediated activation of APJ, with IC50s of 0.70 μM in the cAMP assay, and 1.75 μM in the β-arrestin assay, and EC80 of 10 nM in both assays.

ML221 is a potent apelin (APJ) functional antagonist, inhibiting apelin-13-mediated activation of APJ, with IC50s of 0.70 μM in the cAMP assay, and 1.75 μM in the β-arrestin assay, and EC80 of 10 nM in both assays.

Specifications

Chemical Name or Material ML221
Physical Form Solid
CAS 877636-42-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H11N3O6S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 31 mg/mL (80.45 mM)
SMILES O=C1C=C(CSC2=NC=CC=N2)OC=C1OC(C3=CC=C([N+]([O-])=O)C=C3)=O
Molecular Weight (g/mol) 385.35
Formula Weight 385.35
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.