missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Silver Brand Precision Pipetters

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Individually serialized with Calibration Certificate included.

Brand:  VistaLab™ Technologies 1113

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11905782

  • € 160.00 / Each


Estimated Shipment: 03-09-2021
Add to basket

Description

Description

  • “To deliver” capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Capacity engraved on bonnet

Specifications

Specifications

±1%
1.5 in.
9 in.
1 Ea.
Limited Lifetime
Silver
MLA Precision
400 µL
Not Autoclavable
Nozzle Insert, Calibration Key
±0.5%
Non-sterile
2 in.
Aluminum alloy and Stainless steel
1
Videos
SDS
Documents
Certificates