missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLANA Monoclonal Antibody (OTI6G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807176S
This item is not returnable.
View return policy
Description
MLANA Monoclonal antibody specifically detects MLANA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MLANA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16655 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2315 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G3 | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Human recombinant protein fragment corresponding to amino acids 48-118 of human MLANA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction