Learn More
MLANA Monoclonal Antibody (OTI6G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807176
Encompass_Preferred
 View more versions of this product
Product Code. 17507543
  • € 540.87 / 100µL
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.
Specifications
Specifications
MLANA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16655
MLANA
Human recombit protein fragment corresponding to amino acids 48-118 of human MLANA produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI6G3
Unconjugated
MLANA
MART-1, MART1
Mouse
Affinity Chromatography
RUO
2315
-20° C, Avoid Freeze/Thaw Cycles