missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLANA Mouse anti-Human, Clone: UMAB217, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM870109
This item is not returnable.
View return policy
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.Specifications
| MLANA | |
| Monoclonal | |
| 0.5-1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16655 | |
| MLANA | |
| Human recombit protein fragment corresponding to amino acids 48-118 of human MLANA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| UMAB217 | |
| Unconjugated | |
| MLANA | |
| MART-1, MART1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2315 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction