missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLC1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA537933

Product Code. 15504871

  • € 530.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

MLC1 may be a transporter. It may act as a non-selective neuronal cation channel.The function of this gene product is unknown; however, homology to other proteins suggests that it may be an integral membrane transporter. Mutations in this gene have been associated with megalencephalic leukoencephalopathy with subcortical cysts, an autosomal recessive neurological disorder. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLC1
Polyclonal
Unconjugated
MLC1
AW048630; BB074274; KIAA0027; Kiaa0027-hp; LVM; megalencephalic leukoencephalopathy with subcortical cysts 1; megalencephalic leukoencephalopathy with subcortical cysts 1 gene product; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human); membrane protein MLC1; mKIAA0027; MLC; MLC1; modulator of VRAC current 1; VL; WKL1
Goat
Ammonium sulfate precipitation
RUO
23209
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q15049
MLC1
Peptide with sequence C-SDSANILDEVPFPAR, from the internal region of MLC1 (aa179-193).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.