Learn More
MLC1SA Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99163JF549
Product Code. 18552934
  • € 407.00 / 0.10mL
Description
MLC1SA Polyclonal antibody specifically detects MLC1SA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MLC1SA
Antigen and protein A Affinity-purified