missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLEC Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA532923

Product Code. 13404599

  • € 520.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Platypus (100%).

Carbohydrate-binding protein with a strong ligand preference for Glc2-N-glycan. May play a role in the early steps of protein N-glycosylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLEC
Polyclonal
Unconjugated
Mlec
2410014A08Rik; AI850530; ESTM19; Kiaa0152; malectin; mKIAA0152; Mlec; oligosaccharyltransferase complex subunit (non-catalytic); RGD1307736; X83352
Rabbit
Antigen affinity chromatography
RUO
100050056, 100156249, 100347688, 100755466, 515309, 609274, 9761
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q14165
Mlec
Synthetic 18 amino acid peptide from internal region of human MLEC.
50 μg
Primary
Human, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.