missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLF1 Mouse anti-Human, Clone: UMAB138, lyophilized, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM500100CF
This item is not returnable.
View return policy
Description
MLF1 is involved in lineage commitment of primary hemopoietic progenitors by restricting erythroid formation and enhancing myeloid formation. The protein interferes with erythopoietin-induced erythroid terminal differentiation by preventing cells from exiting the cell cycle through suppression of CDKN1B/p27Kip1 levels. It suppresses RFWD2/COP1 activity via CSN3 which activates p53 and induces cell cycle arrest. It binds DNA and affects the expression of a number of genes so may function as a transcription factor in the nucleus.Specifications
| MLF1 | |
| Monoclonal | |
| Unconjugated | |
| MLF1 | |
| myeloid leukemia factor 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4291 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| UMAB138 | |
| PBS with 8% trehalose and no preservative | |
| P58340 | |
| MLF1 | |
| Full length human recombit protein of human MLF1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction