missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLF2 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504837S
This item is not returnable.
View return policy
Description
MLF2 Monoclonal antibody specifically detects MLF2 in Canine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MLF2 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7TNY6, Q9H3P7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 289312, 611888, 64746 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H2 | |
| Unconjugated | |
| Acbd3 | |
| AI256856; AU018944; C77284; MLF2; Myelodysplasia-myeloid leukemia factor 2; myeloid leukemia factor 2; myeloid leukemia factor 2 homolog; NTN4 | |
| Full length human recombinant protein of human ACBD3 produced in HKE293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction