missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH1 Antibody (164C819), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56552G
This item is not returnable.
View return policy
Description
MLH1 Monoclonal specifically detects MLH1 in Human samples. It is validated for Western Blot.
Specifications
| MLH1 | |
| Monoclonal | |
| DyLight 488 | |
| P40692 | |
| MLH1 | |
| This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 387-403 DSREQKLDAFLQPLSKP of human MLH1; GenBank no. NP_000240.1. | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors | |
| 4292 | |
| Human | |
| Purified |
| Western Blot | |
| 164C819 | |
| Western Blot | |
| COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In Molt-4, a 83 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction