missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH1 Antibody (MLH1/7563), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20598-20ug
This item is not returnable.
View return policy
Description
MLH1 Monoclonal antibody specifically detects MLH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MLH1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1 | |
| Recombinant full-length human MLH1 protein | |
| 20 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors | |
| 4292 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| MLH1/7563 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction