missing translation for 'onlineSavingsMsg'
Learn More

MLH1, Mouse anti-Human, Clone: 4C9C7, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova MAB15272.100uL

Product Code. 16169359

  • € 406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MLH1
Monoclonal
Unconjugated
In ascitic fluid (0.03% sodium azide).
COCA2/FCC2/HNPCC/HNPCC2/MGC5172/hMLH1
Mouse
100 μL
Primary
Human, Monkey
Antibody
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
4C9C7
ELISA (1:10000) Immunofluorescence (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli)
MLH1
Recombinant protein corresponding to amino acids 381-483 of human MLH1.
RUO
4292
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.