missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586455
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
MLH1 is a DNA mismatch repair protein. The repair of mismatch DNA is essential to maintaining the integrity of genetic information over time. An alteration of microsatellite repeats is the result of slippage owing to strand misalignment during DNA replication and is referred to as microsatellite instability (MSI). These defects in DNA repair pathways have been related to human carcinogenesis. The importance of mismatch repair genes became apparent with the identification of the genetic basis for hereditary nonpolyposis colon cancer (HNPC). MSHS2 is involved in the initial cognition of mismatch nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex it is joined by a heterodimer of MLH1 and PMSH, which together help facilitate the later steps in mismatch repair.
Specifications
| MLH1 | |
| Polyclonal | |
| Unconjugated | |
| Mlh1 | |
| 1110035C23Rik; AI317206; AI325952; AI561766; COCA2; colon cancer, nonpolyposis type 2; DNA mismatch repair protein Mlh1; DNA mismatch repair protein Mlh1-like protein; FCC2; hMLH1; HNPCC; HNPCC2; I79_014127; MGC5172; mismatch repair protein 1; MLH1; MLH-1; mutL homolog 1; mutL homolog 1 (E. coli); mutL homolog 1, colon cancer, nonpolyposis type 2; mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli); mutL protein homolog 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P40692 | |
| Mlh1 | |
| Synthetic peptide, corresponding to amino acids 450-500 of Human MLH1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction