missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549933
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MLH3 protein.
This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes. MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination. The protein encoded by this gene functions as a heterodimer with other family members. Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability, and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
Specifications
| MLH3 | |
| Polyclonal | |
| Unconjugated | |
| MLH3 | |
| AV125803; BB126472; CAE; CAE1; cell surface glycoprotein; connexin 50; connexin-50; CTRCT1; CX50; CZP1; DNA mismatch repair protein Mlh3; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; GJA8; HNPCC7; lens fiber protein MP70; lens intrinsic membrane protein MP70; MGC138372; MLH3; MP70; mutL homolog 3; mutL homolog 3 (E coli); MutL protein homolog 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27030 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UHC1 | |
| MLH3 | |
| Synthesized peptide derived from internal of human MLH3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction