missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLH3 Recombinant Rabbit Monoclonal Antibody (6H7J5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA555489

Product Code. 30131087

  • € 436.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen Sequence: QVLQQVDNKF IACLMSTKTE ENGEAGGNLL VLVDQHAAHE RIRLEQLIID SYEKQQAQGS GRKKLLSSTL IPPLEITVTE EQRRLLWCYH KNLEDLGLEF VFPDTSDSLV LVGKVPLCFV EREANELRRG RSTVTKSIVE EFIREQLELL QTTGGIQGTL PLTVQKVLAS QACHGAIKFN DGLSLQESCR LIEALSSCQL.

This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes. MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination. The protein encoded by this gene functions as a heterodimer with other family members. Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability, and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLH3
Recombinant Monoclonal
1.09 mg/mL
PBS with 50% glycerol, 0.05% BSA and 0.09% sodium azide, pH 7.3
Q9UHC1
MLH3
Recombinant protein (or fragment)
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Western Blot
6H7J5
Unconjugated
MLH3
AV125803; BB126472; CAE; CAE1; cell surface glycoprotein; connexin 50; connexin-50; CTRCT1; CX50; CZP1; DNA mismatch repair protein Mlh3; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; GJA8; HNPCC7; lens fiber protein MP70; lens intrinsic membrane protein MP70; MGC138372; MLH3; MP70; mutL homolog 3; mutL homolog 3 (E coli); MutL protein homolog 3
Rabbit
Protein A
RUO
217716, 27030
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.