missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06104MFV610
This item is not returnable.
View return policy
Description
MLK2 Polyclonal antibody specifically detects MLK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MLK2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| EC 2.7.11, MEKK10, mitogen-activated protein kinase kinase kinase 10, Mixed lineage kinase 2, MKN28 kinase, MLK2MKN28 derived nonreceptor_type serine/threonine kinase, MSTEC 2.7.11.25, Protein kinase MST | |
| Produced in rabbits immunized with E. coli-derived Human MLK2 fragment. | |
| 0.1 mL | |
| Apoptosis | |
| 4294 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction