missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19974-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody recognizes the N-term of MAP3K10. The antibody has no cross reaction to MAP3K9.
MLK2 is a member of the serine/threonine kinase family. This kinase has been shown to activate MAPK8/JNK and MKK4/SEK1, and this kinase itself can bephoshorylated, and thus activated by JNK kinases. This kinase functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.Specifications
| MLK2 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K10 | |
| MAP3K10, Mixed lineage kinase 2, MLK2, MST, Protein kinase MST | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4294 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02779 | |
| MAP3K10 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction