missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL, p Ser345 Antibody (JM92-37), Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-66953
This item is not returnable.
View return policy
Description
MLKL Monoclonal specifically detects MLKL in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MLKL (p Ser345) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:100 | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Rabbit | |
| Protein A purified | |
| Necroptosis, Protein Kinase | |
| 197259 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| JM92-37 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| MLKL | |
| Synthetic phospho-peptide corresponding to residues surrounding Ser345 of mouse MLKL aa 314-363/ 472. (SwissProt: Q9D2Y4 Mouse) | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction